Learn More
Research Products International Corp UltraFlux SnapStrip Micro-Tubes, 0.2ml Capacity, Red, 120 Strips per Case
SDP
Catalog No. 50476455
Encompass
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation. 960 tubes per case.
Catalog No. 50-476-455 Supplier Research Products International Corp Supplier No. 145591
UltraFlux thin wall tubes feature precision wall and conical bottom uniformity. This provides optimum tube contact with the temperature block for efficient heat transfer and rapid cycling times. Cap resists opening during thermal cycling and incubation. 960 tubes per case.