Learn More
Alkali Scientific Microtubes, 1.5mL with Green Screw Caps Packed Separately, 500/pk
SDP
Catalog No. 502389571
Encompass_Preferred
These screw cap microtubes come with green caps packed separately. The caps seal tight to prevent evaporation and the tubes can withstand high centrifugal forces.
Catalog No. 50-238-9571 Supplier Alkali Scientific Supplier No. C2310
Add to Cart
Add to Cart
These screw cap microtubes come with green caps packed separately. The caps seal tight to prevent evaporation and the tubes can withstand high centrifugal forces.