Learn More
Andwin Scientific MICROTUBES/1.75ML/2500 CS
SDP
Catalog No. NC0076839
Encompass_Preferred
The 1.7 mL MaxyClear Snaplock Microcentrifuge Tube features great visibility and security with the attached cap that stays closed during centrifugation. The tube is pre-sterilized and is free of RNase, DNase, and pyrogen. It comes in a case of 2500 clear tubes.
Catalog No. NC0076839 Supplier Andwin Scientific Supplier No. MCT175CS
The 1.7 mL MaxyClear Snaplock Microcentrifuge Tube features great visibility and security with the attached cap that stays closed during centrifugation. The tube is pre-sterilized and is free of RNase, DNase, and pyrogen. It comes in a case of 2500 clear tubes.