Learn More
Research Products International Corp Fluorescent Micro-Centrifuge Tubes, Graduated,1.5 ml, 500 per Case
SDP
Catalog No. 50550181
Encompass
Bright colors provide quick sample identification. Polypropylene Tubes feature external graduations, smooth writing surfaces and a convenient target port for rapid sampling. Withstand temperatures from -100°C to +120°C for use in heating blocks or boiling and freezing applications.
Rated to 20,000 x G.
Available with proprietary NoStick resin to ensure maximum sample recovery.
Temperature range: -80°C to +121°C.
Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Tubes are autoclavable and supplied in five assorted fluorescent colors: green, orange, pink, yellow and blue.
Catalog No. 50-550-181 Supplier Research Products International Corp Supplier No. 145551
Bright colors provide quick sample identification. Polypropylene Tubes feature external graduations, smooth writing surfaces and a convenient target port for rapid sampling. Withstand temperatures from -100°C to +120°C for use in heating blocks or boiling and freezing applications.
Rated to 20,000 x G.
Available with proprietary NoStick resin to ensure maximum sample recovery.
Temperature range: -80°C to +121°C.
Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Tubes are autoclavable and supplied in five assorted fluorescent colors: green, orange, pink, yellow and blue.