Learn More
Medchemexpress LLC Microtubule stabilizer-1 | 2699937-76-1 | 99.5% | 390.82 g·mol⁻¹ | C18H17ClF2N6 | 10 MG
SDP
Catalog No. 5000295460
Encompass_Preferred
Microtubule stabilizer-1 is a brain-penetrant small-molecule microtubule stabilizer for preclinical research. It increases acetylated α-tubulin levels and prevents neuronal microtubule loss in toxicity models, supporting studies of Alzheimer's disease and related tauopathies. Supplied as a high-purity powder suitable for laboratory assays and in vivo studies.
  • Brain-penetrant microtubule stabilizer.
  • Increases acetylated α-tubulin levels.
  • Prevents neuronal microtubule loss in toxicity models.
  • Suitable for Alzheimer's disease and tauopathy research.
  • High-purity powder for laboratory use.
  • Available in research pack sizes such as 10 mg.
Catalog No. 50-002-95460 Supplier Medchemexpress LLC Supplier No. HY17654010MG
Microtubule stabilizer-1 is a brain-penetrant small-molecule microtubule stabilizer for preclinical research. It increases acetylated α-tubulin levels and prevents neuronal microtubule loss in toxicity models, supporting studies of Alzheimer's disease and related tauopathies. Supplied as a high-purity powder suitable for laboratory assays and in vivo studies.
  • Brain-penetrant microtubule stabilizer.
  • Increases acetylated α-tubulin levels.
  • Prevents neuronal microtubule loss in toxicity models.
  • Suitable for Alzheimer's disease and tauopathy research.
  • High-purity powder for laboratory use.
  • Available in research pack sizes such as 10 mg.