Learn More
Stoelting™ Microvascular Clip
SDP
Catalog No. 10001365 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
8 mm
11 mm
14 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-001-365 14 mm
10-001-363 8 mm
10-001-364 11 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-001-365 Supplier Stoelting™ Supplier No. 5212004
Add to Cart
Add to Cart
Stoelting™ Microvascular Clips are the most gentle of available Stoelting&Trade; clips. They apply light pressure to make sure no damage is done to the walls of the occluded vessel.
Specifications
For Use With (Application) Neuroscience
Size 14 mm