Learn More
ATCC Microvascular Endothelial Cell Growth Kit-VEGF
Supplier:  ATCC PCS110041
Encompass
Microvascular Endothelial Cell Growth Kit-VEGF contains components that when added to Vascular Cell Basal Medium (ATCC PCS-100-030) create a complete, low serum culture environment for microvascular endothelial cells isolated from normal human tissue.
Catalog No. 50-238-4448