Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Microvascular Growth Supplement (MVGS)

Catalog No. S00525
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00525 2 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00525 Supplier Gibco™ Supplier No. S00525
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Microvascular Growth Supplement (MVGS)

A sterile, concentrated (20X) solution intended for use with Medium 131 to culture human microvascular endothelial cells. Each 25 ml bottle of MVGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human microvascular endothelial cells, and is the correct amount of supplement for a 500 ml bottle of Medium 131. When a 500 ml bottle of medium is supplemented with MVGS, the final concentrations of the components in the supplemented medium are:
  • fetal bovine serum (4.9% v/v final concentration)
  • hydrocortisone (1μg/ml)
  • human fibroblast growth factor (3 ng/ml)
  • heparin (10μg/ml)
  • human epidermal growth factor (1 ng/ml)
  • dibutyryl cyclic AMP (0.08 mM)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage minus 20 degrees C
    Concentration 20 X
    Product Type Growth Supplement
    Sterility Sterile
    Product Line Cascade Biologics
    Quantity 2 Preps
    Shipping Condition Dry Ice
    Species Human
    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.