Learn More
Braintree Scientific Microvette 200, 200 ul,Serum-Gel - Round Bottom, 100/bx
SDP
Catalog No. 501955296
Encompass
Microvette 100/200; Provides all the benefits of a universally suitable capillary blood collection system. The Microvette 100/200 offers the ideal solution for collection of even the smallest blood volumes. Preassembled end-to-end capillary ensures a controlled collection to capillary technique. The Microvette enables blood sampling using either the special rim or gravity-flow principle without any problem. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5296 Supplier Braintree Scientific Supplier No. MV200201291BX
Microvette 100/200; Provides all the benefits of a universally suitable capillary blood collection system. The Microvette 100/200 offers the ideal solution for collection of even the smallest blood volumes. Preassembled end-to-end capillary ensures a controlled collection to capillary technique. The Microvette enables blood sampling using either the special rim or gravity-flow principle without any problem. **Sold for research purposes only and not for human use**