Learn More
Sarstedt Inc Microvette 200 200uL Sample Volume, Conical Inner Tube, 100/Pack
Catalog No. NC9255944 Shop All Sarstedt Inc. Products
Encompass_Preferred
Microvette 200, Serum/clotting activator, 200uL sample volume, Conical inner tube, Special twist cap design is easy to open and minimizes aerosol effect
Catalog No. NC9255944 Supplier Sarstedt Inc Supplier No. 20.1290.100
Microvette 200, Serum/clotting activator, 200uL sample volume, Conical inner tube, Special twist cap design is easy to open and minimizes aerosol effect