Learn More
Braintree Scientific Microvette CB300, 0.3ml Hematology/Potassium EDTA, 100/bx
SDP
Catalog No. NC9694316
Encompass
Microvette CB 300; Capillary collection and sample container in one unit. The Microvette CB 300 capillary tube, along with its attached push cap, is supplied complete with a carrier tube that is also suited for transport and centrifugation. Cylindrical outer tube is ideal for attachment of a barcode label for direct sample analysis. Extremely slim design of the Microvette CB 300's capillary tube ensures optimum serum/plasma recovery following centrifugation. **Sold for research purposes only and not for human use**
Catalog No. NC9694316 Supplier Braintree Scientific Supplier No. MVCB30016444BX
Microvette CB 300; Capillary collection and sample container in one unit. The Microvette CB 300 capillary tube, along with its attached push cap, is supplied complete with a carrier tube that is also suited for transport and centrifugation. Cylindrical outer tube is ideal for attachment of a barcode label for direct sample analysis. Extremely slim design of the Microvette CB 300's capillary tube ensures optimum serum/plasma recovery following centrifugation. **Sold for research purposes only and not for human use**