Learn More
Braintree Scientific MICROVETTE CB 300 W/SERUMUM
SDP
Catalog No. NC0439892
Encompass
Microvette CB 300; Capillary collection and sample container in one unit. The Microvette CB 300 capillary tube, along with its attached push cap, is supplied complete with a carrier tube that is also suited for transport and centrifugation. **Sold for research purposes only and not for human use**
Catalog No. NC0439892 Supplier Braintree Scientific Supplier No. MVCB30016440
Microvette CB 300; Capillary collection and sample container in one unit. The Microvette CB 300 capillary tube, along with its attached push cap, is supplied complete with a carrier tube that is also suited for transport and centrifugation. **Sold for research purposes only and not for human use**