Learn More
Sarstedt Inc MicrovetteCB 300 LH
Supplier:  Sarstedt Inc 16.443.100
Encompass_Preferred
MicrovetteCB 300 l, Lithium Heparin, green US code. Type of collection Capillary - Type of preparation Heparin - Cap: Push cap - Application: Capillary blood collection - Base shape: Flat base -Recommended centrifugation 2000 x g
Catalog No. NC9046728