Learn More
AIR CONTROL INC Microvoid 4F-65-DW VLF Hood
SDP
Catalog No. NC3311174
Encompass_Preferred
The double-walled hood design offers full protection for operators and easy access to the hood interior, while meeting ASHRAE 110-1995, and SEFA 8 requirements. Additional features include Class 100 (optional Class 10) or cleaner Laminar Air Flow, wet process capability, and counterweighted full closure sash . Construction options: standard Type I PVC, polyproyplene, Polypropylene CP7D (FM4910), and Stainless Steel . Safety Features include Full exhaust capability Exhaust failure system, Emergency power off, and Secondary lower tub containment. 4, 6, 8 and 10 feet wide (optional other sizes available), 36 inches deep, and 81 inches high overall. On-board energy efficient motorized impeller/blower(s) to maintain air velocity of 90 Ifm ± 20% with solid state variable speed control. Front removable final HEPA (or optional ULPA) filter, 99.99% efficient on particles 0.3 micron diameter or greater to provide Class 100 laminar flow clean air, MADE IN USA.
Catalog No. NC3311174 Supplier AIR CONTROL INC Supplier No. 4F65TYPE I PVC4FT
Add to Cart
Add to Cart
Request a Quote
The double-walled hood design offers full protection for operators and easy access to the hood interior, while meeting ASHRAE 110-1995, and SEFA 8 requirements. Additional features include Class 100 (optional Class 10) or cleaner Laminar Air Flow, wet process capability, and counterweighted full closure sash . Construction options: standard Type I PVC, polyproyplene, Polypropylene CP7D (FM4910), and Stainless Steel . Safety Features include Full exhaust capability Exhaust failure system, Emergency power off, and Secondary lower tub containment. 4, 6, 8 and 10 feet wide (optional other sizes available), 36 inches deep, and 81 inches high overall. On-board energy efficient motorized impeller/blower(s) to maintain air velocity of 90 Ifm ± 20% with solid state variable speed control. Front removable final HEPA (or optional ULPA) filter, 99.99% efficient on particles 0.3 micron diameter or greater to provide Class 100 laminar flow clean air, MADE IN USA.