Learn More
AIR CONTROL INC Microvoid CS-41 Chemical Hood
SDP
Catalog No. NC3311164
Encompass_Preferred
Features of the Microvoid Model CS-41 include A full unit-width leak containment tub/exhaust plenum, underneath the worksurface, Lip exhaust around all bath or well perimeters for localized high velocity fume containment, Vented base cabinets underneath containment tub/plenum. Construction options: standard Type I PVC, Polypropylene, CP7D (FM4910) polypro, and Stainless Steel . Safety Features include Full exhaust capability Exhaust failure system, Emergency power off, and Secondary lower tub containment. 4, 6, 8 and 10 feet wide (optional other sizes available)A wide array of chemical processing temperature controlled baths, rinse tanks, and process tooling (spinners, hot plate wells, heated ultrasonic cleaning tanks) available for flush/recessed installation within the wet bench. MADE IN USA. Other sizes available.
Catalog No. NC3311164 Supplier AIR CONTROL INC Supplier No. CS41DTYPE I PVC6FT
Add to Cart
Add to Cart
Request a Quote
Features of the Microvoid Model CS-41 include A full unit-width leak containment tub/exhaust plenum, underneath the worksurface, Lip exhaust around all bath or well perimeters for localized high velocity fume containment, Vented base cabinets underneath containment tub/plenum. Construction options: standard Type I PVC, Polypropylene, CP7D (FM4910) polypro, and Stainless Steel . Safety Features include Full exhaust capability Exhaust failure system, Emergency power off, and Secondary lower tub containment. 4, 6, 8 and 10 feet wide (optional other sizes available)A wide array of chemical processing temperature controlled baths, rinse tanks, and process tooling (spinners, hot plate wells, heated ultrasonic cleaning tanks) available for flush/recessed installation within the wet bench. MADE IN USA. Other sizes available.