Learn More
AIR CONTROL INC MicrovoidCS-50D High Eff Hood
SDP
Catalog No. NC3311168
Encompass_Preferred
Revolutionary Microvoid CS-50D Wet Process hood utilizing the Air Sentry High Performance technology saving 33-40% in exhausted air required to safely operate, vs.traditional wet process stations. Constructed in UL-1805 compliant Type I PVC, the wet station has full width leak containment tub/exhaust plenum underneath the worksurface. A wide array of chemical processing heating baths, rinse tanks, and process tooling (spinners, hot plate wells, etc. are available for flush/recessed installation within the wet bench. Lip exhaust around all bath or well perimeters for localized high velocity fume containment. Complete PLC/HMI interface and control of all wet bench system parameters traditionally available in all Microvoid Wet Process Station models. All safety systems and interlocks/inhibits, alarm hierarchies, and BMS communications capabilities available in all Microvoid Wet Process Stations. MADE IN USA. Other sizes and materials of construction available.
Catalog No. NC3311168 Supplier AIR CONTROL INC Supplier No. CD50DTYPE I PVC6FT
Add to Cart
Add to Cart
Request a Quote
Revolutionary Microvoid CS-50D Wet Process hood utilizing the Air Sentry High Performance technology saving 33-40% in exhausted air required to safely operate, vs.traditional wet process stations. Constructed in UL-1805 compliant Type I PVC, the wet station has full width leak containment tub/exhaust plenum underneath the worksurface. A wide array of chemical processing heating baths, rinse tanks, and process tooling (spinners, hot plate wells, etc. are available for flush/recessed installation within the wet bench. Lip exhaust around all bath or well perimeters for localized high velocity fume containment. Complete PLC/HMI interface and control of all wet bench system parameters traditionally available in all Microvoid Wet Process Station models. All safety systems and interlocks/inhibits, alarm hierarchies, and BMS communications capabilities available in all Microvoid Wet Process Stations. MADE IN USA. Other sizes and materials of construction available.