Learn More
AIR CONTROL INC MicrovoidModel 4F-55 VLF Hood
SDP
Catalog No. NC3311158
Encompass_Preferred
The Type I PVC material hood offers full protection for operators and easy access to the hood interior, while meeting ASHRAE 110-1995, and SEFA 8 requirements. Additional features include Class 100 (optional Class 10) or cleaner Laminar Air Flow, wet process capability, and counterweighted full closure sash . Construction options: standard Type I PVC, Polypropylene, CP7D polypropylene (FM4910), and Stainless Steel . Safety Features include Full exhaust capability Exhaust failure system, Emergency power off, and Secondary lower tub containment. 4, 6, 8 and 10 feet wide (optional other sizes available), 36-in deep, and 81-in high overall. On-board energy efficient motorized impeller/blower(s) to maintain air velocity of 90 Ifm /20% with solid state variable speed control. Front removable final HEPA (or optional ULPA) filter, 99.99% efficient on particles 0.3 micron diameter or greater to provide Class 100 laminar flow clean air. MADE IN USA. Other sizes available.
Catalog No. NC3311158 Supplier AIR CONTROL INC Supplier No. 4F55TYPE I PVC6FT
Add to Cart
Add to Cart
Request a Quote
The Type I PVC material hood offers full protection for operators and easy access to the hood interior, while meeting ASHRAE 110-1995, and SEFA 8 requirements. Additional features include Class 100 (optional Class 10) or cleaner Laminar Air Flow, wet process capability, and counterweighted full closure sash . Construction options: standard Type I PVC, Polypropylene, CP7D polypropylene (FM4910), and Stainless Steel . Safety Features include Full exhaust capability Exhaust failure system, Emergency power off, and Secondary lower tub containment. 4, 6, 8 and 10 feet wide (optional other sizes available), 36-in deep, and 81-in high overall. On-board energy efficient motorized impeller/blower(s) to maintain air velocity of 90 Ifm /20% with solid state variable speed control. Front removable final HEPA (or optional ULPA) filter, 99.99% efficient on particles 0.3 micron diameter or greater to provide Class 100 laminar flow clean air. MADE IN USA. Other sizes available.