Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Microvolume Pipet Tips, Sterile
GSA_VAGREENER_CHOICE

Catalog No. 07200521 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 10 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
07-200-521 0.1 to 10 μL
07-200-524 0.5 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-200-521 Supplier Corning™ Supplier No. 4894
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide accurate, reliable performance in the stated volume range

  • Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
  • RNase-/DNase-free.
  • Nonpyrogenic.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.1 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Certifications/Compliance EN ISO 13485 compliant, FDA CFR 21 Part 820 compliant (cGMP), BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, DNase/RNase Free, Quality Control Tested
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Microvolume, Round
Packaging Rack
Quantity 96/Pk., 960/Cs.
Product Type Pipette Tips
Includes Case, Racks, Tips
For Use With (Equipment) Gilson Pipetman, Universal Fit Ultramicro
Graduated Yes
Material Polypropylene
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.