Learn More
Quidel Corporation MicroVue CH50 Eq EIA C1 through C9
SDP
Supplier:  Quidel Corporation A018
Encompass
The MicroVue CH50 Eq EIA measures the total classical complement pathway activity in human serum and allows detection of a deficiency of one or more of the complement components C1 through C9.
Catalog No. NC0279543