Learn More
Quidel Corporation MicroVue CIC-Raji Cell Replacement Enzyme Immunoassay
SDP
Catalog No. NC1925216 Shop All QuidelOrtho Products
Encompass
The MicroVue CIC-Raji Cell Replacement Enzyme Immunoassay measures immune complexes containing C3 activation fragments in human serum and plasma.
This item is not returnable. View return policy
Catalog No. NC1925216 Supplier Quidel Corporation Supplier No. A002
The MicroVue CIC-Raji Cell Replacement Enzyme Immunoassay measures immune complexes containing C3 activation fragments in human serum and plasma.