Learn More
Quidel Corporation Micro Vue Creatinine assay Kit;
Supplier: Quidel Corporation 8009
The MicroVue Creatinine assay is a quantitative, colorimetric assay based on a modified Jaffe method where alkaline picrate forms a colored solution in the presence of creatinine.