Learn More
Quidel Corporation MICROVUE SC5B-9 PLUS EIA
SDP
Supplier:  Quidel Corporation A020
Encompass
The MicroVue SC5b-9 Plus Enzyme Immunoassay measures the amount of the SC5b-9 complex in specimens.
Catalog No. NC9713652