Learn More
Electron Microscopy Sciences Microwave Calibration Slide Set
SDP
Catalog No. 5027284
Encompass_Preferred
A calibration slide for microwave staining. A set which includes two glass slides. Slide #1 has liquid crystal squares corresponding to the following temperatures: 35 degree C, 40 degree C, 45 degree C; Slide #2 has liquid crystal squares corresponding to the following temperatures: 50 degree C, 55 degree C, and 60 degree C.
This calibration set will ensure reproducible programming of the oven to achieve an ideal target temperature of large staining volumes (50-200 ml solution). Developed in collaboration with Dr. Gary Login, Beth Israel Hospital, Boston, MA
Catalog No. 50-272-84 Supplier Electron Microscopy Sciences Supplier No. 9703101
A calibration slide for microwave staining. A set which includes two glass slides. Slide #1 has liquid crystal squares corresponding to the following temperatures: 35 degree C, 40 degree C, 45 degree C; Slide #2 has liquid crystal squares corresponding to the following temperatures: 50 degree C, 55 degree C, and 60 degree C.
This calibration set will ensure reproducible programming of the oven to achieve an ideal target temperature of large staining volumes (50-200 ml solution). Developed in collaboration with Dr. Gary Login, Beth Israel Hospital, Boston, MA