Learn More
Electron Microscopy Sciences EMS Microwave Non-Vacuum Processor Kit
SDP
Catalog No. 501926680
Encompass_Preferred
For processing of tissue samples in the microwave processor. The non vacuum kit includes Non Vacuum Processor Bowl, Processor Tray, 74 Cassette Processing Rack
Catalog No. 50-192-6680 Supplier Electron Microscopy Sciences Supplier No. 9705212
For processing of tissue samples in the microwave processor. The non vacuum kit includes Non Vacuum Processor Bowl, Processor Tray, 74 Cassette Processing Rack