Learn More
Electron Microscopy Sciences EMS MPC9000 Microwave Processing Chamber
SDP
Catalog No. 501901847
Encompass_Preferred
MPC 9000 processing chamber, developed only for use the EMS 9000 laboratory microwave, provides consistent processing, vacuum infiltration, and quick specimen turnaround. The sturdy and chemical resistant guide allows exact placement of the chamber. This minimizes inconsistent results caused by different microwave distribution patterns within the microwaves chamber.
The chamber uses standard EMS Lynx I & II specimen vials that lock into the tip-proof stations. Processing baskets can be mixed to match the workload. Feed-through for process temperature probe placement while under vacuum. Low maintenance chamber disassembles for easy cleaning.
Catalog No. 50-190-1847 Supplier Electron Microscopy Sciences Supplier No. 97045
MPC 9000 processing chamber, developed only for use the EMS 9000 laboratory microwave, provides consistent processing, vacuum infiltration, and quick specimen turnaround. The sturdy and chemical resistant guide allows exact placement of the chamber. This minimizes inconsistent results caused by different microwave distribution patterns within the microwaves chamber.
The chamber uses standard EMS Lynx I & II specimen vials that lock into the tip-proof stations. Processing baskets can be mixed to match the workload. Feed-through for process temperature probe placement while under vacuum. Low maintenance chamber disassembles for easy cleaning.