Learn More
Electron Microscopy Sciences Microwave Slide Staining Holder with Disp Tray
SDP
Catalog No. 5030571
Encompass
Slide staining holder with disposible tray is for use in laboratory microwave oven. Each rack holds 24 slides.
Catalog No. 50-305-71 Supplier Electron Microscopy Sciences Supplier No. 97090
Slide staining holder with disposible tray is for use in laboratory microwave oven. Each rack holds 24 slides.