Learn More
Electron Microscopy Sciences EMS Microwave Vacuum Processor Kit
SDP
Catalog No. 501926678
Encompass_Preferred
For processing of tissue samples in the microwave processor. Complete kit includes vacuum processor bowl, processor tray, vacuum processor tubing kit, and 74 cassette processing rack.
Catalog No. 50-192-6678 Supplier Electron Microscopy Sciences Supplier No. 9705210
For processing of tissue samples in the microwave processor. Complete kit includes vacuum processor bowl, processor tray, vacuum processor tubing kit, and 74 cassette processing rack.