Learn More
Sigma Aldrich Fine Chemicals Biosciences Nunc-Immuno MicroWell 96 well solid plates, 60/cs
Catalog No. NC1162061
Encompass
Nunc-Immuno 96 MicroWell solid plates are intended for quantitative and qualitative solid phase immuno assays (i.e. ELISA) and binding assays.
Catalog No. NC1162061 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M57851CS
Nunc-Immuno 96 MicroWell solid plates are intended for quantitative and qualitative solid phase immuno assays (i.e. ELISA) and binding assays.