Learn More
Biosurface Technologies Corp MicroWell Biofilm Plate Assay, for Rapid Biofilm Testing, Annodized Aluminum, fits most standard 48-well plates.
Catalog No. NC3662431
Encompass
"The MicroWell™ Biofilm Plate Assay holds 48 coupons, each measuring 0.25 inches by 0.80 inches, inside the wells. This versatility allows researchers to choose from a wide variety of materials, including custom options upon request, to tailor their studies to specific needs.Sterility and convenience are at the forefront of the MicroWell™ assay’s design. The system utilizes the disposable lid that comes with 48-well plates, fitting securely over the coupon holder plate to prevent contamination during incubation. Additionally, the anodized aluminum plate is fully autoclavable, allowing for the reuse of the system. This feature ensures long-term reliability and promotes cost-efficiency and environmental sustainability by reducing the need for disposable components.Designed to facilitate biofilm assays similar to the Calgary Biofilm Device using ASTM E2799*, the MicroWell™ system offers expanded material choices.
Catalog No. NC3662431 Supplier Biosurface Technologies Corp Supplier No. MBPA 48
"The MicroWell™ Biofilm Plate Assay holds 48 coupons, each measuring 0.25 inches by 0.80 inches, inside the wells. This versatility allows researchers to choose from a wide variety of materials, including custom options upon request, to tailor their studies to specific needs.Sterility and convenience are at the forefront of the MicroWell™ assay’s design. The system utilizes the disposable lid that comes with 48-well plates, fitting securely over the coupon holder plate to prevent contamination during incubation. Additionally, the anodized aluminum plate is fully autoclavable, allowing for the reuse of the system. This feature ensures long-term reliability and promotes cost-efficiency and environmental sustainability by reducing the need for disposable components.Designed to facilitate biofilm assays similar to the Calgary Biofilm Device using ASTM E2799*, the MicroWell™ system offers expanded material choices.