Learn More
Medchemexpress LLC MID-1 100mg | 312608-54-1 | 293.30 g·mol-1 | C12H11N3O4S | 100 MG
SDP
Catalog No. 5000226485
Encompass_Preferred
MID-1 (Benzamide, 4-ethoxy-N-(5-nitro-2-thiazolyl); CAS 312608-54-1) is a small-molecule disruptor of the MG53-IRS-1 interaction that increases IRS-1 protein levels and enhances insulin signaling in vitro. It is supplied as a solid and as a 10 mM solution in DMSO for research use.
  • Disrupts MG53-IRS-1 protein-protein interaction in vitro.
  • Increases IRS-1 protein level and insulin signaling.
  • Available as solid and as 10 mM DMSO solution.
  • High purity (≈99.4%).
Catalog No. 50-002-26485 Supplier Medchemexpress LLC Supplier No. HY115461100MG
MID-1 (Benzamide, 4-ethoxy-N-(5-nitro-2-thiazolyl); CAS 312608-54-1) is a small-molecule disruptor of the MG53-IRS-1 interaction that increases IRS-1 protein levels and enhances insulin signaling in vitro. It is supplied as a solid and as a 10 mM solution in DMSO for research use.
  • Disrupts MG53-IRS-1 protein-protein interaction in vitro.
  • Increases IRS-1 protein level and insulin signaling.
  • Available as solid and as 10 mM DMSO solution.
  • High purity (≈99.4%).