Learn More
Medchemexpress LLC MID-1 25mg | 312608-54-1 | 25 MG
SDP
Catalog No. 5000229400
Encompass_Preferred
MID-1 is a small-molecule research compound that disrupts the interaction between MG53 and IRS-1, preventing MG53-induced ubiquitination and degradation of IRS-1 and thereby increasing IRS-1 levels and insulin signaling in muscle cells. Supplied as an orange to red solid for laboratory research.
  • Disrupts MG53-IRS-1 interaction
  • Enhances IRS-1 expression and insulin signaling
  • High purity for reliable assay performance
  • Orange to red solid with good DMSO solubility
  • Suitable for in vitro studies of insulin signaling and glucose uptake
Catalog No. 50-002-29400 Supplier Medchemexpress LLC Supplier No. HY11546125MG
MID-1 is a small-molecule research compound that disrupts the interaction between MG53 and IRS-1, preventing MG53-induced ubiquitination and degradation of IRS-1 and thereby increasing IRS-1 levels and insulin signaling in muscle cells. Supplied as an orange to red solid for laboratory research.
  • Disrupts MG53-IRS-1 interaction
  • Enhances IRS-1 expression and insulin signaling
  • High purity for reliable assay performance
  • Orange to red solid with good DMSO solubility
  • Suitable for in vitro studies of insulin signaling and glucose uptake