Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MID-1 50mg | 312608-54-1 | 50 MG
SDP

Catalog No. 5000228919
Encompass_Preferred

MID-1 is a small-molecule research compound reported to disrupt the interaction between MG53 and IRS-1, thereby modulating insulin signaling. Supplied for laboratory research use, the compound includes manufacturer-provided data on purity, solubility, and storage for preclinical experiments.

  • High purity: 99.36%.
  • Molecular weight: 293.30 g/mol.
  • Chemical formula: C12H11N3O4S.
  • Physical state: solid, orange to red.
  • Soluble in DMSO: 31.25 mg/mL (sonication recommended).
  • Recommended storage: powder at -20°C (up to 3 years); in solvent at -80°C (up to 6 months).
  • Available packaging sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.

Catalog No. 50-002-28919 Supplier Medchemexpress LLC Supplier No. HY11546150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MID-1 is a small-molecule research compound reported to disrupt the interaction between MG53 and IRS-1, thereby modulating insulin signaling. Supplied for laboratory research use, the compound includes manufacturer-provided data on purity, solubility, and storage for preclinical experiments.

  • High purity: 99.36%.
  • Molecular weight: 293.30 g/mol.
  • Chemical formula: C12H11N3O4S.
  • Physical state: solid, orange to red.
  • Soluble in DMSO: 31.25 mg/mL (sonication recommended).
  • Recommended storage: powder at -20°C (up to 3 years); in solvent at -80°C (up to 6 months).
  • Available packaging sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.