Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K1 Series Mid-Sole Ice Cleat

Catalog No. 19060831
Encompass
Encompass_Preferred
Change view
Click to view available options
Style:
Low Profile Tungston Carbide Molded in
Original Profile Tungston Carbide Molded in
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
19-060-831 Original Profile Tungston Carbide Molded in
19-060-832 Low Profile Tungston Carbide Molded in
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-060-831 Supplier K1 Series Supplier No. V9770150O/S
Add to Cart
Add to Cart
This item is not returnable. View return policy

The K1 Series Mid-Sole ice cleat is the one-size-fits-all winter traction solution. The ice-penetrating tungsten carbide spikes maximize grip while the rotatable design provides seamless transition when traction is not required.

  • Available in Original, Low-Profile, and High-Profile styles to provide near-universal coverage of all footwear
  • Strategically placed ice-penetrating tungsten carbide spikes provide maximum traction on ice and snow
  • Easy to use design rotates to top of foot to allow driving, climbing, and indoor use
  • Black or hi-viz adjustable strap delivers a perfect fit regardless of shoe size
  • Specially formulated rubber pad provides maximum comfort and durability even in extreme cold weather conditions

Specifications

Closure Type Elastic
Color Black
For Use With (Application) Outdoor Surfaces, Ice Surfaces
Size Universal
Strap Material Elastic
Style Original Profile Tungston Carbide Molded in
Disposable Reusable
Gender Unisex
Waterproof Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.