Learn More
Ace Glass, Inc. MID-STAND RECTANGLE PLATE
SDP
Catalog No. 5000854729
Encompass
The Ace kilo-scale modular stand is a mobile platform for reactors and filtration vessels from 10 liters on up to 150 liters. Modular design allows a customer's existing stand to be modified to accept 200mm,300mm,400mm and 450mm flanged vessels as well as accommodating rod mounted or flange mounted stirring motors via interchangeable parts. Carriage lift is operated via a ratcheting handle or converted to an electric motor lift with the optional 12852-670 kit.
Catalog No. 50-008-54729 Supplier Ace Glass, Inc. Supplier No. 12852640
The Ace kilo-scale modular stand is a mobile platform for reactors and filtration vessels from 10 liters on up to 150 liters. Modular design allows a customer's existing stand to be modified to accept 200mm,300mm,400mm and 450mm flanged vessels as well as accommodating rod mounted or flange mounted stirring motors via interchangeable parts. Carriage lift is operated via a ratcheting handle or converted to an electric motor lift with the optional 12852-670 kit.