Learn More
MSC Milwaukee Tool Pipeline Inspection Reel, 120 ft., Mid-Stiff, Self-Leveling, 1 ea.
Catalog No. 5000510595
Encompass
The 120 ft. Mid-Stiff Modular Pipeline Inspection Reel is designed for professional plumbing and diagnostic applications, supporting inspection of lines ranging from two to six inches. This reel features a 25 mm self-leveling HDR camera head with a sapphire crystal lens for clear imaging. It integrates with the 500GB control hub to provide high-resolution video and picture capture, distance tracking, and sonde frequency monitoring.
  • Self-leveling HDR camera head for accurate image orientation
  • Compatible with the control hub for advanced recording and job management
  • Includes small and medium camera head skids for varied pipe diameters
  • Pitch sensing capability and integrated sonde tracking frequencies
  • Rugged design with removable backpack straps for job site portability
  • Sapphire crystal lens provides durability and clarity
  • Digital zoom function up to 4X
Catalog No. 50-005-10595 Supplier MSC Supplier No. 57159154
The 120 ft. Mid-Stiff Modular Pipeline Inspection Reel is designed for professional plumbing and diagnostic applications, supporting inspection of lines ranging from two to six inches. This reel features a 25 mm self-leveling HDR camera head with a sapphire crystal lens for clear imaging. It integrates with the 500GB control hub to provide high-resolution video and picture capture, distance tracking, and sonde frequency monitoring.
  • Self-leveling HDR camera head for accurate image orientation
  • Compatible with the control hub for advanced recording and job management
  • Includes small and medium camera head skids for varied pipe diameters
  • Pitch sensing capability and integrated sonde tracking frequencies
  • Rugged design with removable backpack straps for job site portability
  • Sapphire crystal lens provides durability and clarity
  • Digital zoom function up to 4X