Learn More
TARGETMOL CHEMICALS INC MIDAGLIZOLE HYDROCHLORID 5MG
SDP
Catalog No. 502581599
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Midaglizole hydrochloride ((±)-DG5128) (DG5128) is a preferred (alpha)2-adrenoceptor antagonist. Midazolazole hydrochloride (DG5128) has an affinity for (alpha)2-adrenoceptor (pKi = 6.28) 7.4 times higher than that of (alpha)1-adrenoceptor. purity: 98%
Catalog No. 50-258-1599 Supplier TARGETMOL CHEMICALS INC Supplier No. T110155MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Midaglizole hydrochloride ((±)-DG5128) (DG5128) is a preferred (alpha)2-adrenoceptor antagonist. Midazolazole hydrochloride (DG5128) has an affinity for (alpha)2-adrenoceptor (pKi = 6.28) 7.4 times higher than that of (alpha)1-adrenoceptor. purity: 98%