Learn More
Medchemexpress LLC Midaglizole (hydrochloride) | 79689-25-1 | 324.25 | 100 MG
SDP
Catalog No. 5000448565
Encompass_Preferred
Midaglizole hydrochloride is a potent alpha2-adrenoceptor antagonist. It is a hypoglycemic agent that increases blood pressure and reduces blood glucose levels in vivo.
  • Potent alpha2-adrenoceptor antagonist
  • Hypoglycemic agent
  • Increases blood pressure in vivo
  • Reduces blood glucose levels in vivo
Catalog No. 50-004-48565 Supplier Medchemexpress LLC Supplier No. HYU00165100MG
Midaglizole hydrochloride is a potent alpha2-adrenoceptor antagonist. It is a hypoglycemic agent that increases blood pressure and reduces blood glucose levels in vivo.
  • Potent alpha2-adrenoceptor antagonist
  • Hypoglycemic agent
  • Increases blood pressure in vivo
  • Reduces blood glucose levels in vivo