Learn More
Medchemexpress LLC Midaglizole hydrochloride | 79689-25-1 | 324.25 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYU001655MG
Encompass_Preferred
Midaglizole hydrochloride is a potent α2-adrenoceptor antagonist and a hypoglycemic agent. It has been shown to increase blood pressure and reduce blood glucose levels in vivo.
  • Potent α2-adrenoceptor antagonist
  • Hypoglycemic agent
  • Increases blood pressure in rats
  • Reduces blood glucose levels in dogs
  • Stimulates insulin release in rat islets and MIN6 B-cell line
Catalog No. 50-004-48499