Learn More
Medchemexpress LLC Midaglizole hydrochloride | 79689-25-1 | 324.25 | 50 MG
SDP
Catalog No. 5000448563
Encompass_Preferred
Midaglizole hydrochloride, also known as (±)-DG5128 or DG5128, is a potent α2-adrenoceptor antagonist and a hypoglycemic agent. It has been shown to increase blood pressure and reduce blood glucose levels in vivo. It stimulates insulin release with EC50 values of 200 nM for rat islets and 24 μM for MIN6 B-cell lines.
  • Acts as a potent α2-adrenoceptor antagonist
  • Functions as a hypoglycemic agent
  • Increases blood pressure in rats in vivo
  • Reduces blood glucose levels in dogs in vivo
  • Stimulates insulin release from pancreatic islets
Catalog No. 50-004-48563 Supplier Medchemexpress LLC Supplier No. HYU0016550MG
Midaglizole hydrochloride, also known as (±)-DG5128 or DG5128, is a potent α2-adrenoceptor antagonist and a hypoglycemic agent. It has been shown to increase blood pressure and reduce blood glucose levels in vivo. It stimulates insulin release with EC50 values of 200 nM for rat islets and 24 μM for MIN6 B-cell lines.
  • Acts as a potent α2-adrenoceptor antagonist
  • Functions as a hypoglycemic agent
  • Increases blood pressure in rats in vivo
  • Reduces blood glucose levels in dogs in vivo
  • Stimulates insulin release from pancreatic islets