Learn More
Med Vet International Midamox For Cats, 9.1 To 18 Pounds, Purple Label, 6 Dose
Catalog No. 502828850
Encompass_Preferred
Midamox for Cats (imidacloprid/moxidectin) is indicated for the prevention of heartworm disease caused by Dirofilaria immiti in cats. Midamox for Cats kills adult fleas (Ctenocephalides felis) and is indicated for the treatment of fleas infestations. Midamox for Cats is also indicated for the treatment and control of ear mite (Otodectes cynotis) infestations and the following parasites: hookworm species (Ancylostoma tubaeforme) and roundworm species (Toxocara cati).
Catalog No. 50-282-8850 Supplier Med Vet International Supplier No. RXMIDAMOXCPRV
Midamox for Cats (imidacloprid/moxidectin) is indicated for the prevention of heartworm disease caused by Dirofilaria immiti in cats. Midamox for Cats kills adult fleas (Ctenocephalides felis) and is indicated for the treatment of fleas infestations. Midamox for Cats is also indicated for the treatment and control of ear mite (Otodectes cynotis) infestations and the following parasites: hookworm species (Ancylostoma tubaeforme) and roundworm species (Toxocara cati).