Learn More
Synthware™ Middle Adapter/ Pump Adapter for Vacuum Trap Body
GREENER_CHOICESDP
Catalog No. 31501800 Shop All Kemtech Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
31-501-800 1 Ea.
1 options
Catalog No. 31-501-800 Supplier Synthware™ Supplier No. V175038
Add to Cart
Add to Cart
Spherical joints add flexibility to the Vacuum Trap.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Product Type Middle/Pump Adapter
Quantity 1 Ea.
Standard Taper Joint No. 35/20
Width (Metric) 12 mm