Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Middle School Earth Science Bulletin Board Set
SDPSCI_ED

Catalog No. p-4230914
Change view
Click to view available options
Product Type:
Extreme Climate and Weather Bulletin Board Set
Subject:
Social Science
Grade:
4 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Subject Grade
S02413 Extreme Climate and Weather Bulletin Board Set Social Science 4 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02413 Supplier Carson Dellosa Publishing Supplier No. CD410022

An informative earth science themed bulletin board.

  • Supports NSE and NCTM standards

Specifications

Grade 4 to 8
Format Set
Product Type Extreme Climate and Weather Bulletin Board Set
Includes 5 Charts (17 x 24 in.) with realistic artwork and facts about deserts, tropical rain forests, hurricanes, prevailing winds and tornadoes, a resource guide
Subject Social Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.