Learn More
Fisher Science Education™ Middle School Series Stereo Microscope
SCI_ED
Catalog No. s72593 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification Power:
2X, 4X
Eyepiece:
WF 10X
Electrical Requirements:
Rechargeable, cordless
Catalog No. Magnification Power Eyepiece Electrical Requirements
S72593 2X, 4X WF 10X Rechargeable, cordless
1 options
Catalog No. S72593
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Stereoscopic Microscope for Macro-Size Specimens is ideal for middle school classrooms
Optical System
  • 10/15X WF eyepiece
  • Dual magnification
Viewing Head
  • Binocular viewing head with 45° incline
  • Interpupillary adjustment of 55mm to 75mm
  • Diopter Adjustment
Focusing
  • Coarse focus adjustment
  • 40mm height adjustment range on focusing mechanism
  • 70mm working distance
Stand
  • Low profile plain stage with clips
  • Pole stand to accommodate larger sample
  • Compact design
Illumination
  • Eco-friendly and energy efficient LED illumination
  • LED ring in transmitted light, provides homogeneous light distribution
  • LED incident Illumination
  • Membrane switch controls
Warranty and Services
Limited Lifetime Warranty
Specifications
Includes Eyepieces, Stand, Dust Cover, Objectives
Voltage 110/220 V
Magnification Power 2X, 4X
Electrical Requirements Rechargeable, cordless
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Product Type Microscope
Eyepiece WF 10X
For the specifics on a particular model, see the specification table located at the bottom of the page of that individual model