Learn More
MOLTOX Molecular Toxicology Inc Middlebrook 7H10 w/ OADC Enrichment, 10 mL/tube, 10 tubes/box Store in Refrigerator
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 22105
Encompass_Preferred
Middlebrook 7H10 Agar is used in conjunction with OADC Enrichment for the isolation and cultivation of Mycobacteria. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-707