Learn More
Medchemexpress LLC HY-B1908 50mg Medchemexpress, Midecamycin CAS:35457-80-8 Purity:>98%
SDP
Catalog No. 501933036
Encompass_Preferred
Medchemexpress, HY-B1908 50mg Midecamycin CAS:35457-80-8 Midecamycin, an acetoxy-substituted macrolide antibiotic, is tested against gram-positive and gram-negative bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3036 Supplier Medchemexpress LLC Supplier No. HYB190850MG
Medchemexpress, HY-B1908 50mg Midecamycin CAS:35457-80-8 Midecamycin, an acetoxy-substituted macrolide antibiotic, is tested against gram-positive and gram-negative bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.