Learn More
Ace Glass, Inc. Midget bubbler, Complete with 0-25mL (5mL divisions)graduated bottle and #7 Ace-Safe™; stopper with porosity A (145-174 micron) filter tip
SDP
Catalog No. 50974998
Encompass_Preferred
Midget bubbler, Complete with 0-25mL (5mL divisions)graduated bottle and #7 Ace-Safe™; stopper with porosity A (145-174 micron) filter tip
Catalog No. 50-974-998 Supplier Ace Glass, Inc. Supplier No. 753318
Midget bubbler, Complete with 0-25mL (5mL divisions)graduated bottle and #7 Ace-Safe™; stopper with porosity A (145-174 micron) filter tip