Learn More
Ace Glass, Inc. Midget bubbler complete with 0-30mL bottle and stopper
SDP
Catalog No. 509557830
Encompass_Preferred
0-30mL midget bubbler complete with stopper and an impinger bottle with an open tube end inlet/outlet and a sintered glass filter with porosity A. Bottle has encircling graduations from 0-30mL in 5mL divisions. Used in determination of Acrolein: 4-Hexylresorcinol Method #1.
Catalog No. 50-955-7830 Supplier Ace Glass, Inc. Supplier No. 753220
0-30mL midget bubbler complete with stopper and an impinger bottle with an open tube end inlet/outlet and a sintered glass filter with porosity A. Bottle has encircling graduations from 0-30mL in 5mL divisions. Used in determination of Acrolein: 4-Hexylresorcinol Method #1.