Learn More
DELTA SCIENTIFIC MIDI CENTRIFGE TUBE,5ML,10BAGS
Catalog No. NC2267063
Encompass
Diamond Midi Centrifuge Tube, 5.0mL, PP, Assembled Blue Screw Cap, Graduated, STERILE, 20/Bag, 10 Bags/Unit
This item is not returnable. View return policy
Catalog No. NC2267063 Supplier DELTA SCIENTIFIC Supplier No. 111580BS
Diamond Midi Centrifuge Tube, 5.0mL, PP, Assembled Blue Screw Cap, Graduated, STERILE, 20/Bag, 10 Bags/Unit