Learn More
Hoefer, Inc Hoefer™ SUB13 Midi Submarine Gel Electrophoresis Unit
SDP
Catalog No. NC2741103
Encompass_Preferred
Ideal for analytical and preparative studies of nucleic acidsFeatures and BenefitsColor-coded and height adjustable combs—easily identify comb thickness and control well depthBuffer recirculation ports—may be connected to a peristaltic pump for buffer recirculation during electrophoresis to maintain buffer pH and prevent ionic gradient formationCompact tank—reduces the buffer volume required to cover the gel, providing greater control over the voltage gradient and run-timeUV-transparent running traySide handles—for safe and easy transportationIncludes:Buffer Chamber Assembly; Safety Lid; High Voltage Leads; Running Tray with Silicone Gasket; 16-well combs, 2 pcs; Colored Loading Strips; Buffer Recirculation PortsSpecifications:Gel Dimensions (w x l): 12.8 x 15 cmMaximum Buffer Volume: 900 mlMaximum Power Settings: 300 V, 200 mA, 60 WMaximum Sample Capacity: 112Unit Dimensions (w x l x h): 20 x 32 x 7 cmSafety Certifications: EN61010-1, CE
Catalog No. NC2741103 Supplier Hoefer, Inc Supplier No. SUB13
Ideal for analytical and preparative studies of nucleic acidsFeatures and BenefitsColor-coded and height adjustable combs—easily identify comb thickness and control well depthBuffer recirculation ports—may be connected to a peristaltic pump for buffer recirculation during electrophoresis to maintain buffer pH and prevent ionic gradient formationCompact tank—reduces the buffer volume required to cover the gel, providing greater control over the voltage gradient and run-timeUV-transparent running traySide handles—for safe and easy transportationIncludes:Buffer Chamber Assembly; Safety Lid; High Voltage Leads; Running Tray with Silicone Gasket; 16-well combs, 2 pcs; Colored Loading Strips; Buffer Recirculation PortsSpecifications:Gel Dimensions (w x l): 12.8 x 15 cmMaximum Buffer Volume: 900 mlMaximum Power Settings: 300 V, 200 mA, 60 WMaximum Sample Capacity: 112Unit Dimensions (w x l x h): 20 x 32 x 7 cmSafety Certifications: EN61010-1, CE