Learn More
Alkali Scientific Microflex MidKnight MK-296 Distinctive, Black Nitrile Exam Gloves, 1000/Case
SDP
Catalog No. 502336073
Encompass_Preferred
The MidKnight™ MK-296 Black Nitrile Exam Gloves boast exceptional durability, and their deep hue effectively conceals dirt, oil, and grease, ensuring a consistently polished and professional appearance for workers' hands. Beyond stain concealment, the dark color provides excellent contrast, aiding workers in identifying lighter materials during handling. These disposable grip gloves are latex-free, fully textured, and non-foaming, making them suitable for individuals with latex allergies. Whether in wet laboratory or industrial settings, these robust gloves deliver a secure grip. The MidKnight™ MK-296 Black Nitrile Exam Gloves meet medical examination standards, comply with U.S. Food Contact regulations, and are powder-free for added convenience. Packaged at 100 gloves per box, 1000 gloves per case.
Catalog No. 50-233-6073 Supplier Alkali Scientific Supplier No. NG2920
The MidKnight™ MK-296 Black Nitrile Exam Gloves boast exceptional durability, and their deep hue effectively conceals dirt, oil, and grease, ensuring a consistently polished and professional appearance for workers' hands. Beyond stain concealment, the dark color provides excellent contrast, aiding workers in identifying lighter materials during handling. These disposable grip gloves are latex-free, fully textured, and non-foaming, making them suitable for individuals with latex allergies. Whether in wet laboratory or industrial settings, these robust gloves deliver a secure grip. The MidKnight™ MK-296 Black Nitrile Exam Gloves meet medical examination standards, comply with U.S. Food Contact regulations, and are powder-free for added convenience. Packaged at 100 gloves per box, 1000 gloves per case.